Assay Details
Gene Symbol: ZDHHC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039617.1 | NP_001034706.1 | 3 - 4 | 523 | 67 | |
NR_135617.1 | 3 - 4 | 531 | 67 | |||
NR_135618.1 | 3 - 4 | 531 | 67 | |||
XM_005269279.2 | XP_005269336.1 | 3 - 4 | 531 | 67 | ||
XM_006713493.2 | XP_006713556.1 | 3 - 4 | 501 | 67 | ||
XM_006713494.2 | XP_006713557.1 | 3 - 4 | 429 | 67 | ||
XM_011512417.1 | XP_011510719.1 | 2 - 3 | 288 | 67 | ||
GenBank mRNA | AK122907.1 | - | 3 - 4 | 531 | 67 | |
BC022078.2 | - | 3 - 4 | 523 | 4827355 | 67 | |
BG721568.1 | - | 3 - 4 | 528 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03002156.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03038126.hg.1 | TC03002156.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03038126.hg.1 | ENST00000296326.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03038126.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03038126.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300202284.hg.1 | TC0300013696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000296326.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000397544.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000438232.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000460442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000465519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000488508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | ENST00000497422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013696.hg.1 | NM_001039617.1 | Refseq |
