Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145243.4 | NP_660286.1 | 5 - 6 | 1124 | 111 | |
| GenBank mRNA | AB048348.1 | - | 5 - 6 | 1107 | 111 | |
| AK091101.1 | - | 4 - 5 | 535 | 111 | ||
| BC012915.1 | - | 5 - 6 | 1105 | 4471592 | 111 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006360.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
diet induced thermogenesis
glucose metabolic process
misfolded or incompletely synthesized protein catabolic process
lipid metabolic process
response to stress
negative regulation of mitochondrial fusion
mitochondrial protein processing
cristae formation
energy homeostasis
metalloendopeptidase activity
metal ion binding
diet induced thermogenesis
glucose metabolic process
misfolded or incompletely synthesized protein catabolic process
lipid metabolic process
response to stress
negative regulation of mitochondrial fusion
mitochondrial protein processing
cristae formation
energy homeostasis
metalloendopeptidase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000460671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000460671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000482274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000482274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | ENST00000482274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042492.hg.1 | TC01006360.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042492.hg.1 | ENST00000358603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042492.hg.1 | ENST00000371226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042492.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042495.hg.1 | TC01006360.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042495.hg.1 | ENST00000358603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042495.hg.1 | ENST00000371226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042495.hg.1 | ENST00000469854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01042495.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242217.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242217.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242217.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242217.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242217.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242217.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242217.hg.1 | TC0100018446.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100242215.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242215.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242215.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242215.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242215.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242215.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242215.hg.1 | TC0100018446.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000371226.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000419242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000421528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000426139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000453710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000456980.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000460671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000460671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000467509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000476933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000482274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000482274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | ENST00000482274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018446.hg.1 | NM_145243.4 | Refseq |

OMA1
Genomic Map