Assay Details
| Gene Symbol: STOML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145286.2 | NP_660329.1 | 1 - 2 | 190 | 94 | |
| GenBank mRNA | AK313632.1 | - | 1 - 2 | 162 | 94 | |
| BC025760.1 | - | 1 - 2 | 177 | 5204252 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000583.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313632.1,BC025760.1,NM_145286.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13006695.hg.1 | ENST00000379631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006695.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006695.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006695.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006695.hg.1 | TC13000583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006693.hg.1 | ENST00000379631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006693.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006693.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006693.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006693.hg.1 | TC13000583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158504.hg.1 | TC1300008659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008659.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158500.hg.1 | TC1300008659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | ENST00000379631.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | ENST00000423210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | NM_001144033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | NM_145286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300158503.hg.1 | TC1300008659.hg.1 | Clariom D TC |

STOML3
Genomic Map