Assay Details
| Gene Symbol: KIFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145754.3 | NP_665697.1 | 7 - 8 | 893 | 105 | |
| XM_005272357.3 | XP_005272414.1 | 7 - 8 | 983 | 105 | ||
| XM_011517362.2 | XP_011515664.1 | 7 - 8 | 984 | 105 | ||
| GenBank mRNA | AK055045.1 | - | 3 - 5 | 105 | ||
| BC017311.2 | - | 7 - 9 | 5090622 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000854.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK055045.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08011393.hg.1 | TC08000854.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011393.hg.1 | ENST00000301331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011393.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011393.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163467.hg.1 | TC0800009262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |

KIFC2
Genomic Map