Assay Details
| Gene Symbol: TRAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145803.2 | NP_665802.1 | 1 - 2 | 267 | 97 | |
| GenBank mRNA | BC031052.1 | - | 1 - 2 | 212 | 5272008 | 97 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001560.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC031052.1,NM_145803.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
activation of MAPK activity
protein polyubiquitination
ossification
neural tube closure
stimulatory C-type lectin receptor signaling pathway
toll-like receptor signaling pathway
positive regulation of T cell cytokine production
MyD88-dependent toll-like receptor signaling pathway
MyD88-independent toll-like receptor signaling pathway
protein complex assembly
I-kappaB kinase/NF-kappaB signaling
activation of NF-kappaB-inducing kinase activity
JNK cascade
antigen processing and presentation of exogenous peptide antigen via MHC class II
osteoclast differentiation
membrane protein intracellular domain proteolysis
positive regulation of protein ubiquitination
positive regulation of lipopolysaccharide-mediated signaling pathway
activation of protein kinase activity
positive regulation of interleukin-2 production
toll-like receptor 9 signaling pathway
Fc-epsilon receptor signaling pathway
T-helper 1 type immune response
positive regulation of T cell proliferation
odontogenesis of dentin-containing tooth
myeloid dendritic cell differentiation
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interleukin-12 biosynthetic process
positive regulation of interleukin-6 biosynthetic process
bone resorption
positive regulation of osteoclast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell development
positive regulation of smooth muscle cell proliferation
T cell receptor signaling pathway
positive regulation of T cell activation
regulation of immunoglobulin secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
protein autoubiquitination
nucleotide-binding oligomerization domain containing signaling pathway
interleukin-1-mediated signaling pathway
protein K63-linked ubiquitination
response to interleukin-1
cellular response to lipopolysaccharide
positive regulation of transcription regulatory region DNA binding
ubiquitin-protein transferase activity
tumor necrosis factor receptor binding
protein binding
zinc ion binding
ligase activity
protein kinase binding
mitogen-activated protein kinase kinase kinase binding
ubiquitin conjugating enzyme binding
ubiquitin protein ligase binding
thioesterase binding
identical protein binding
histone deacetylase binding
protein kinase B binding
protein N-terminus binding
ubiquitin protein ligase activity
negative regulation of transcription from RNA polymerase II promoter
activation of MAPK activity
protein polyubiquitination
ossification
neural tube closure
stimulatory C-type lectin receptor signaling pathway
toll-like receptor signaling pathway
positive regulation of T cell cytokine production
MyD88-dependent toll-like receptor signaling pathway
MyD88-independent toll-like receptor signaling pathway
protein complex assembly
I-kappaB kinase/NF-kappaB signaling
activation of NF-kappaB-inducing kinase activity
JNK cascade
antigen processing and presentation of exogenous peptide antigen via MHC class II
osteoclast differentiation
membrane protein intracellular domain proteolysis
positive regulation of protein ubiquitination
positive regulation of lipopolysaccharide-mediated signaling pathway
activation of protein kinase activity
positive regulation of interleukin-2 production
toll-like receptor 9 signaling pathway
Fc-epsilon receptor signaling pathway
T-helper 1 type immune response
positive regulation of T cell proliferation
odontogenesis of dentin-containing tooth
myeloid dendritic cell differentiation
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interleukin-12 biosynthetic process
positive regulation of interleukin-6 biosynthetic process
bone resorption
positive regulation of osteoclast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell development
positive regulation of smooth muscle cell proliferation
T cell receptor signaling pathway
positive regulation of T cell activation
regulation of immunoglobulin secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
protein autoubiquitination
nucleotide-binding oligomerization domain containing signaling pathway
interleukin-1-mediated signaling pathway
protein K63-linked ubiquitination
response to interleukin-1
cellular response to lipopolysaccharide
positive regulation of transcription regulatory region DNA binding
ubiquitin-protein transferase activity
tumor necrosis factor receptor binding
protein binding
zinc ion binding
ligase activity
protein kinase binding
mitogen-activated protein kinase kinase kinase binding
ubiquitin conjugating enzyme binding
ubiquitin protein ligase binding
thioesterase binding
identical protein binding
histone deacetylase binding
protein kinase B binding
protein N-terminus binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019583.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019583.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019583.hg.1 | TC11001560.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100180090.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180090.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180090.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180090.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180090.hg.1 | TC1100010551.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000348124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000526995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | ENST00000529150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_004620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010551.hg.1 | NM_145803.2 | Refseq |

TRAF6
Genomic Map