Assay Details
| Gene Symbol: NUMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005745.1 | NP_001005745.1 | 5 - 6 | 518 | 80 | |
| NM_003744.5 | NP_003735.3 | 5 - 6 | 518 | 80 | ||
| GenBank mRNA | AF171940.1 | - | 5 - 6 | 468 | 80 | |
| AF171941.1 | - | 5 - 6 | 468 | 80 | ||
| AK315130.1 | - | 4 - 5 | 399 | 80 | ||
| BC020788.1 | - | 4 - 5 | 399 | 80 | ||
| BC068476.1 | - | 5 - 6 | 518 | 4822300 | 80 | |
| BX248781.1 | - | 4 - 5 | 413 | 80 | ||
| EU265735.1 | - | 3 - 4 | 237 | 80 | ||
| EU265737.1 | - | 3 - 4 | 237 | 80 | ||
More Information
Set Membership: |
|

NUMB
Genomic Map