Assay Details
| Gene Symbol: MAGEA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005366.4 | NP_005357.2 | 2 - 3 | 194 | 63 | |
| XM_017029521.1 | XP_016885010.1 | 4 - 5 | 405 | 63 | ||
| GenBank mRNA | AY747607.1 | - | 2 - 3 | 152 | 63 | |
| BC004479.2 | - | 2 - 3 | 130 | 3833322 | 63 | |
| BQ212898.1 | - | 2 - 3 | 194 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000713.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008691.hg.1 | ENST00000424803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009158.hg.1 | TC0X000713.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009158.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009158.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009160.hg.1 | TC0X000713.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009160.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009160.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009162.hg.1 | TC0X000713.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009162.hg.1 | ENST00000333104.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009162.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009162.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009162.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158901.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158901.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158901.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158901.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158901.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158901.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158901.hg.1 | TC0X00008689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008691.hg.1 | ENST00000424803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158903.hg.1 | TC0X00008689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00158898.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158898.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158898.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158898.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158898.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158898.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158898.hg.1 | TC0X00008689.hg.1 | Clariom D TC |

MAGEA11
Genomic Map