Assay Details
| Gene Symbol: DEPDC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145208.1 | NP_001138680.1 | 3 - 4 | 521 | 79 | |
| NM_018369.2 | NP_060839.2 | 3 - 4 | 521 | 79 | ||
| XM_011543509.2 | XP_011541811.1 | 3 - 4 | 514 | 79 | ||
| XM_011543510.2 | XP_011541812.1 | 3 - 4 | 489 | 79 | ||
| XM_017009629.1 | XP_016865118.1 | 3 - 4 | 489 | 79 | ||
| GenBank mRNA | AF303178.1 | - | 3 - 4 | 489 | 79 | |
| AF490257.1 | - | 3 - 4 | 448 | 79 | ||
| AK002114.1 | - | 3 - 4 | 489 | 79 | ||
| AK290684.1 | - | 3 - 4 | 516 | 79 | ||
| AK299242.1 | - | 4 - 5 | 555 | 79 | ||
| AK300782.1 | - | 3 - 4 | 489 | 79 | ||
| BC019075.1 | - | 3 - 4 | 484 | 5091386 | 79 | |
| DC299445.1 | - | 3 - 4 | 522 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001392.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000509006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000509006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019444.hg.1 | TC05001392.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019444.hg.1 | ENST00000265036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019444.hg.1 | ENST00000453022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019444.hg.1 | ENST00000545085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019444.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019444.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175741.hg.1 | TC0500010865.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000265036.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000453022.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000505017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000509006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000509006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512078.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | ENST00000512452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_001145208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010865.hg.1 | NM_018369.2 | Refseq |

DEPDC1B
Genomic Map