Assay Details
| Gene Symbol: DKFZP434K028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026882.1 | 3 - 4 | 518 | 102 | ||
| GenBank mRNA | BC021187.1 | - | 3 - 4 | 509 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001856.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P327679 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P327679 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 15103 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 2945 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P327679 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P327679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240539 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829275 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16458148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16458150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16458154 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240541 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240527 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240527 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16458148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16458150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829275 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16458154 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240539 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28240541 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07829263 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P327679 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02454359 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P327679 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 4630424 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 4630424 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P327679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458138 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458154 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829250 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829196 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829272 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829208 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829184 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829266 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829216 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458148 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458112 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829230 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458116 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458156 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829282 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829248 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829213 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829269 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458160 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829201 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829275 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458158 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829259 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829239 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829279 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829247 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829186 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829254 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829265 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829273 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458114 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829278 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458140 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829182 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16458157 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829221 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829189 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07829246 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7948602 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7948603 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7948604 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7948605 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7948601 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7948600 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7948599 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1554880_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 234356_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021446.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021446.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021446.hg.1 | TC11001856.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100204192.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204192.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204192.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204192.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204192.hg.1 | TC1100013169.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013169.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204190.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204190.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204190.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204190.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204190.hg.1 | TC1100013169.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100204191.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204191.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204191.hg.1 | ENST00000244906.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204191.hg.1 | NR_026882.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204191.hg.1 | TC1100013169.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 234356_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1554880_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1554880_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234356_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554880_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234356_at8 | - | - |
| Human Genome U133 Set | 234356_at | - | - |
| Human Genome U133 Set | 234356_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045253 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045253 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 224193 | - | - |

DKFZP434K028
Genomic Map