Assay Details
| Gene Symbol: RRP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015056.2 | NP_055871.1 | 10 - 11 | 1100 | 83 | |
| GenBank mRNA | AK056263.1 | - | 10 - 11 | 1063 | 83 | |
| BC014005.2 | - | 10 - 11 | 1072 | 83 | ||
| BC028386.2 | - | 10 - 11 | 1105 | 4821672 | 83 | |
| D80001.1 | - | 10 - 11 | 998 | 83 | ||
More Information
Set Membership: |
|

RRP1B
Genomic Map