Assay Details
| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182491.2 | NP_872297.2 | 2 - 3 | 357 | 149 | |
| XM_017012774.1 | XP_016868263.1 | 2 - 3 | 357 | 149 | ||
| GenBank mRNA | AK027118.1 | - | 2 - 3 | 299 | 149 | |
| AK290170.1 | - | 2 - 3 | 316 | 149 | ||
| BC016963.1 | - | 2 - 3 | 292 | 149 | ||
| BC029558.1 | - | 2 - 3 | 300 | 5263788 | 149 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001080.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290170.1,BC029558.1,NM_182491.2,XM_017012774.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07016583.hg.1 | TC07001080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016583.hg.1 | ENST00000316495.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016583.hg.1 | ENST00000401903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016583.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171176.hg.1 | TC0700010005.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000471448.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000478137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | ENST00000574135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010005.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000316495.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000397083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000401903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | ENST00000484977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | NM_182491.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700171178.hg.1 | TC0700010005.hg.1 | Clariom D TC |

ZFAND2A
Genomic Map