Assay Details
| Gene Symbol: ZNF295-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027273.2 | 1 - 2 | 162 | 68 | ||
| GenBank mRNA | BC029588.1 | - | 1 - 2 | 162 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000196.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21001946.hg.1 | ENST00000455701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001946.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001946.hg.1 | TC21000196.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001947.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001947.hg.1 | TC21000196.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000455701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000455701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000455701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119385.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119385.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119385.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007235.hg.1 | NR_119385.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001948.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001948.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001948.hg.1 | TC21000196.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100148783.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148783.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148783.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148783.hg.1 | TC2100007235.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148783.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148783.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148783.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000412906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000429739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000455701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000455701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000455701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | ENST00000596595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_027273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119384.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119385.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119385.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119385.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007235.hg.1 | NR_119385.1 | Refseq |

ZNF295-AS1
Genomic Map