Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203467.1 | NP_982292.1 | 2 - 3 | 597 | 94 | |
| GenBank mRNA | BC030142.2 | - | 2 - 3 | 533 | 94 | |
| BC093697.1 | - | 2 - 3 | 299 | 94 | ||
| BC107888.1 | - | 2 - 3 | 346 | 94 | ||
| BC112241.1 | - | 2 - 3 | 299 | 94 | ||
More Information
Set Membership: |
|

LRR1
Genomic Map