Assay Details
| Gene Symbol: TTC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138376.2 | NP_612385.2 | 7 - 8 | 899 | 88 | |
| GenBank mRNA | AK074553.1 | - | 7 - 8 | 882 | 88 | |
| BC008647.1 | - | 7 - 8 | 840 | 88 | ||
| BC030822.1 | - | 7 - 8 | 843 | 4243419 | 88 | |
| BC053538.1 | - | 7 - 8 | 847 | 5225152 | 88 | |
| BX248066.1 | - | 7 - 8 | 671 | 88 | ||
| BX640732.1 | - | 7 - 8 | 778 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000877.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14009520.hg.1 | ENST00000258821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009520.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009520.hg.1 | TC14000877.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000553828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000553828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000553828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400181246.hg.1 | TC1400010709.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000258821.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000383029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000553828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000553828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000553828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000554157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | ENST00000557379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010709.hg.1 | NM_138376.2 | Refseq |

TTC5
Genomic Map