Assay Details
| Gene Symbol: CENPU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024629.3 | NP_078905.2 | 8 - 9 | 866 | 107 | |
| NR_104593.1 | 8 - 9 | 866 | 107 | |||
| XM_005263218.4 | XP_005263275.2 | 8 - 9 | 901 | 107 | ||
| GenBank mRNA | AF469667.1 | - | 8 - 9 | 869 | 107 | |
| AF516710.1 | - | 9 - 10 | 1283 | 107 | ||
| AK027121.1 | - | 6 - 7 | 676 | 107 | ||
| AK292297.1 | - | 8 - 9 | 812 | 107 | ||
| BC031520.1 | - | 3 - 4 | 214 | 107 | ||
| BC067802.1 | - | 8 - 9 | 806 | 107 | ||
| BC107744.1 | - | 3 - 4 | 311 | 107 | ||
| BC131556.1 | - | 8 - 9 | 866 | 40108113 | 107 | |
| BC141854.1 | - | 8 - 9 | 864 | 8860370 | 107 | |
| CR457376.1 | - | 1 - 2 | 70 | 107 | ||
| DQ907910.1 | - | 8 - 9 | 796 | 107 | ||
| DQ907911.1 | - | 8 - 9 | 796 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001808.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04024639.hg.1 | TC04001808.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024639.hg.1 | ENST00000281453.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024639.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024639.hg.1 | ENST00000541665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024639.hg.1 | ENST00000541971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024639.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186370.hg.1 | TC0400012620.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000281453.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000502461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000506535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000508095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000510146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | ENST00000514781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NM_024629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012620.hg.1 | NR_104593.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 147135 | - | - |

CENPU
Genomic Map