Assay Details
| Gene Symbol: AIFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018060.2 | NP_001018070.1 | 4 - 5 | 571 | 113 | |
| NM_001146288.1 | NP_001139760.1 | 4 - 5 | 656 | 113 | ||
| NM_144704.2 | NP_653305.1 | 4 - 5 | 571 | 113 | ||
| GenBank mRNA | AK055035.1 | - | 4 - 5 | 555 | 113 | |
| AK094844.1 | - | 4 - 5 | 560 | 113 | ||
| BC032485.1 | - | 4 - 5 | 538 | 5221203 | 113 | |
| CR456342.1 | - | 3 - 4 | 390 | 113 | ||
More Information
Set Membership: |
|

AIFM3
Genomic Map