Assay Details
| Gene Symbol: REEP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005669.4 | NP_005660.4 | 1 - 2 | 258 | 72 | |
| XM_017009844.1 | XP_016865333.1 | 1 - 2 | 175 | 72 | ||
| GenBank mRNA | AK295436.1 | - | 1 - 2 | 149 | 72 | |
| AK298268.1 | - | 1 - 2 | 162 | 72 | ||
| AK299902.1 | - | 1 - 2 | 154 | 72 | ||
| AK300048.1 | - | 1 - 2 | 163 | 72 | ||
| AK312522.1 | - | 1 - 2 | 163 | 72 | ||
| AY562243.1 | - | 1 - 2 | 114 | 72 | ||
| BC000232.1 | - | 1 - 2 | 139 | 3350749 | 72 | |
| BC065926.1 | - | 1 - 2 | 117 | 6164621 | 72 | |
| BX648005.1 | - | 1 - 2 | 239 | 72 | ||
| DA807043.1 | - | 1 - 2 | 259 | 72 | ||
| M74090.1 | - | 1 - 2 | 140 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001678.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05022757.hg.1 | TC05001678.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022757.hg.1 | ENST00000379638.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022757.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022757.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022757.hg.1 | ENST00000513339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022757.hg.1 | ENST00000545426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022757.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022761.hg.1 | TC05001678.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022761.hg.1 | ENST00000379638.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022761.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022761.hg.1 | ENST00000513339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022761.hg.1 | ENST00000545426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022761.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022766.hg.1 | TC05001678.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022766.hg.1 | ENST00000379638.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022766.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022765.hg.1 | TC05001678.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022765.hg.1 | ENST00000379638.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022765.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022765.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011721.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181055.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181055.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181055.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181055.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181055.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181055.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181055.hg.1 | TC0500011721.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500181056.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181056.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181056.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181056.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181056.hg.1 | TC0500011721.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000261482.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000474542.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000497856.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181046.hg.1 | TC0500011721.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000379638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000504247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000511865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | ENST00000513339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | NM_005669.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181050.hg.1 | TC0500011721.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 110183 | - | - |

REEP5
Genomic Map