Assay Details
| Gene Symbol: IFI35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005533.4 | NP_005524.2 | 5 - 6 | 792 | 66 | |
| XM_005257302.4 | XP_005257359.1 | 5 - 6 | 771 | 66 | ||
| XM_017024584.1 | XP_016880073.1 | 5 - 6 | 764 | 66 | ||
| XM_017024585.1 | XP_016880074.1 | 5 - 6 | 749 | 66 | ||
| GenBank mRNA | AK025406.1 | - | 3 - 4 | 387 | 66 | |
| BC001356.2 | - | 5 - 6 | 709 | 3050452 | 66 | |
| BM050205.1 | - | 4 - 5 | 67 | |||
| U72882.1 | - | 4 - 5 | 604 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000545.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17006643.hg.1 | ENST00000415816.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006643.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006643.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006643.hg.1 | TC17000545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006642.hg.1 | ENST00000415816.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006642.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006642.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006642.hg.1 | TC17000545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000396722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000396722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000396722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000536969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000536969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000536969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157975.hg.1 | TC1700007931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157979.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157979.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157979.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157979.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157979.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157979.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157979.hg.1 | TC1700007931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157976.hg.1 | TC1700007931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000246911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000396722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000396722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000396722.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000415816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000438323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000534876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000536969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000536969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000536969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000538473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | ENST00000546325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007931.hg.1 | NM_005533.4 | Refseq |

IFI35
Genomic Map