Assay Details
| Gene Symbol: TIMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138379.2 | NP_612388.2 | 4 - 5 | 818 | 77 | |
| XM_011534694.2 | XP_011532996.1 | 4 - 5 | 789 | 77 | ||
| GenBank mRNA | AK312746.1 | - | 4 - 5 | 788 | 77 | |
| BC008988.1 | - | 4 - 5 | 818 | 4184237 | 77 | |
| JX049980.1 | - | 4 - 5 | 761 | 77 | ||
| KU178832.1 | - | 4 - 5 | 761 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001976.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05027519.hg.1 | TC05001976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027519.hg.1 | ENST00000274532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027519.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027520.hg.1 | TC05001976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027520.hg.1 | ENST00000274532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027520.hg.1 | ENST00000407087.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027520.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027520.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187823.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187823.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187823.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187823.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187823.hg.1 | TC0500012587.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187824.hg.1 | TC0500012587.hg.1 | Clariom D TC |

TIMD4
Genomic Map