Assay Details
| Gene Symbol: TIMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138379.2 | NP_612388.2 | 5 - 6 | 897 | 67 | |
| GenBank mRNA | AK312746.1 | - | 5 - 6 | 867 | 67 | |
| BC008988.1 | - | 5 - 6 | 897 | 4184237 | 67 | |
| JX049980.1 | - | 5 - 6 | 840 | 67 | ||
| KU178832.1 | - | 5 - 6 | 840 | 67 | ||
| KU178833.1 | - | 4 - 5 | 759 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001976.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027516.hg.1 | TC05001976.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027516.hg.1 | ENST00000274532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027516.hg.1 | ENST00000406964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027516.hg.1 | ENST00000407087.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027516.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027516.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012587.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | ENST00000274532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | ENST00000406964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | ENST00000407087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | NM_001146726.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | NM_138379.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187821.hg.1 | TC0500012587.hg.1 | Clariom D TC |

TIMD4
Genomic Map