Assay Details
| Gene Symbol: FGF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002009.3 | NP_002000.1 | 1 - 2 | 260 | 85 | |
| GenBank mRNA | BC010956.1 | - | 1 - 2 | 201 | 4282925 | 85 |
| DB206350.1 | - | 1 - 2 | 343 | 85 | ||
| M60828.1 | - | 1 - 2 | 176 | 85 | ||
| S81661.1 | - | 1 - 2 | 176 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000386.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC010956.1,M60828.1,NM_002009.3,S81661.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
ovarian follicle development
positive regulation of defense response to virus by host
phosphatidylcholine biosynthetic process
signal transduction
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
epidermis development
response to wounding
mesenchymal cell proliferation
positive regulation of keratinocyte proliferation
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
hair follicle morphogenesis
actin cytoskeleton reorganization
protein localization to cell surface
phosphatidylinositol-3-phosphate biosynthetic process
wound healing
surfactant homeostasis
positive regulation of GTPase activity
positive regulation of transcription, DNA-templated
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of keratinocyte migration
positive regulation of cell division
branching involved in salivary gland morphogenesis
positive regulation of epithelial cell proliferation involved in lung morphogenesis
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
secretion by lung epithelial cell involved in lung growth
xenophagy
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
type 2 fibroblast growth factor receptor binding
protein binding
growth factor activity
heparin binding
1-phosphatidylinositol-3-kinase activity
chemoattractant activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MAPK cascade
ovarian follicle development
positive regulation of defense response to virus by host
phosphatidylcholine biosynthetic process
signal transduction
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
epidermis development
response to wounding
mesenchymal cell proliferation
positive regulation of keratinocyte proliferation
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
hair follicle morphogenesis
actin cytoskeleton reorganization
protein localization to cell surface
phosphatidylinositol-3-phosphate biosynthetic process
wound healing
surfactant homeostasis
positive regulation of GTPase activity
positive regulation of transcription, DNA-templated
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of keratinocyte migration
positive regulation of cell division
branching involved in salivary gland morphogenesis
positive regulation of epithelial cell proliferation involved in lung morphogenesis
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
secretion by lung epithelial cell involved in lung growth
xenophagy
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
type 2 fibroblast growth factor receptor binding
protein binding
growth factor activity
heparin binding
1-phosphatidylinositol-3-kinase activity
chemoattractant activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15003590.hg.1 | ENST00000267843.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003590.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003590.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003590.hg.1 | TC15000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003587.hg.1 | ENST00000267843.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003587.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003587.hg.1 | TC15000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003589.hg.1 | ENST00000267843.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003589.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003589.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003589.hg.1 | TC15000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151224.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151224.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151224.hg.1 | TC1500007185.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151220.hg.1 | TC1500007185.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000560979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007185.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151221.hg.1 | TC1500007185.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500151216.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151216.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151216.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151216.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151216.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151216.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151216.hg.1 | TC1500007185.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000267843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000560270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000560765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | ENST00000614567.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | NM_002009.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151222.hg.1 | TC1500007185.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 150104 | - | - |

FGF7
Genomic Map