Assay Details
| Gene Symbol: SERPINE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006216.3 | NP_006207.1 | 6 - 7 | 1226 | 139 | |
| XM_005246641.2 | XP_005246698.1 | 6 - 7 | 1315 | 139 | ||
| XM_017004329.1 | XP_016859818.1 | 6 - 7 | 1207 | 139 | ||
| XM_017004330.1 | XP_016859819.1 | 6 - 7 | 1044 | 139 | ||
| GenBank mRNA | AK312499.1 | - | 6 - 7 | 1240 | 139 | |
| BC015663.2 | - | 6 - 7 | 1183 | 4578406 | 139 | |
More Information
Set Membership: |
|

SERPINE2
Genomic Map