| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000522619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000522619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000522619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006036.hg.1 |
TC08000435.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006036.hg.1 |
ENST00000276570.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006036.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006036.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006037.hg.1 |
TC08000435.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006037.hg.1 |
ENST00000276570.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006037.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006037.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006038.hg.1 |
TC08000435.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006038.hg.1 |
ENST00000276570.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006038.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08006038.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153960.hg.1 |
TC0800007846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800153962.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153962.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153962.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153962.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153962.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153962.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153962.hg.1 |
TC0800007846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000522619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000522619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000522619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007846.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
ENST00000276570.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
ENST00000519330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
ENST00000524076.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
NM_033105.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153961.hg.1 |
TC0800007846.hg.1 |
Clariom D TC |