| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008456.hg.1 |
TC05000608.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008456.hg.1 |
ENST00000296666.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008456.hg.1 |
ENST00000330542.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008456.hg.1 |
ENST00000442138.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008456.hg.1 |
ENST00000512535.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008456.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008456.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000512871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500158726.hg.1 |
TC0500008558.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000296666.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000442138.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000507774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000512871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
ENST00000513427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_001286808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008558.hg.1 |
NM_130809.4 |
Refseq |