Assay Details
| Gene Symbol: BRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242786.1 | NP_001229715.1 | 3 - 4 | 511 | 100 | |
| NM_001242787.1 | NP_001229716.1 | 3 - 4 | 511 | 100 | ||
| NM_001242788.1 | NP_001229717.1 | 2 - 3 | 721 | 100 | ||
| NM_001519.3 | NP_001510.2 | 3 - 4 | 802 | 100 | ||
| XM_005267561.3 | XP_005267618.1 | 3 - 4 | 824 | 100 | ||
| XM_011536672.2 | XP_011534974.1 | 3 - 4 | 824 | 100 | ||
| XM_011536673.1 | XP_011534975.1 | 3 - 4 | 824 | 100 | ||
| GenBank mRNA | AK294899.1 | - | 3 - 4 | 511 | 100 | |
| AK295579.1 | - | 3 - 4 | 424 | 100 | ||
| BC086856.1 | - | 2 - 3 | 454 | 6166751 | 100 | |
| U28838.1 | - | 3 - 4 | 805 | 100 | ||
| U75276.1 | - | 3 - 4 | 506 | 100 | ||
More Information
Set Membership: |
|

BRF1
Genomic Map