Assay Details
| Gene Symbol: TRANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014831.2 | NP_055646.2 | 19 - 20 | 5638 | 97 | |
| XM_006713432.3 | XP_006713495.1 | 20 - 21 | 5806 | 97 | ||
| XM_011534289.2 | XP_011532591.1 | 20 - 21 | 6567 | 97 | ||
| XM_017007570.1 | XP_016863059.1 | 19 - 20 | 5438 | 97 | ||
| XM_017007571.1 | XP_016863060.1 | 20 - 21 | 5699 | 97 | ||
| XM_017007572.1 | XP_016863061.1 | 20 - 21 | 5601 | 97 | ||
| GenBank mRNA | AB002340.3 | - | 9 - 10 | 4016 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001283.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03022003.hg.1 | TC03001283.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022003.hg.1 | ENST00000301807.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022003.hg.1 | ENST00000428977.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022003.hg.1 | ENST00000429976.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022003.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300177923.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300177923.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300177923.hg.1 | ENST00000429976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300177923.hg.1 | NM_014831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300177923.hg.1 | TC0300010714.hg.1 | Clariom D TC |

TRANK1
Genomic Map