Assay Details
| Gene Symbol: URB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014825.2 | NP_055640.2 | 27 - 28 | 4707 | 89 | |
| GenBank mRNA | AB011111.2 | - | 22 - 23 | 3941 | 89 | |
| AF231919.1 | - | 27 - 28 | 4545 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000397.hg.1
Set Membership: |
|

URB1
Genomic Map