Assay Details
| Gene Symbol: MAST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015016.1 | NP_055831.1 | 25 - 26 | 3211 | 81 | |
| XM_005259827.2 | XP_005259884.1 | 24 - 25 | 3229 | 81 | ||
| XM_006722700.3 | XP_006722763.1 | 24 - 25 | 3371 | 81 | ||
| XM_017026509.1 | XP_016881998.1 | 25 - 26 | 3562 | 81 | ||
| XM_017026510.1 | XP_016881999.1 | 24 - 25 | 3542 | 81 | ||
| XM_017026516.1 | XP_016882005.1 | 25 - 26 | 3559 | 81 | ||
| GenBank mRNA | AB011133.1 | - | 25 - 26 | 3209 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000351.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004652.hg.1 | ENST00000262811.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004652.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004652.hg.1 | TC19000351.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004653.hg.1 | ENST00000262811.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004653.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004653.hg.1 | TC19000351.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010001.hg.1 | ENST00000600364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010001.hg.1 | ENST00000600364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153969.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153969.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153969.hg.1 | TC1900007363.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153969.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153969.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000601226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000608648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | ENST00000609494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007363.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010001.hg.1 | ENST00000600364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010001.hg.1 | ENST00000600364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153967.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153967.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153967.hg.1 | TC1900007363.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153967.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153967.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153968.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153968.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153968.hg.1 | TC1900007363.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153968.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153968.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153965.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153965.hg.1 | NM_015016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153965.hg.1 | TC1900007363.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153965.hg.1 | ENST00000262811.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153965.hg.1 | NM_015016.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 146806 | - | - |

MAST3
Genomic Map