| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
TC02002264.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
ENST00000263710.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
ENST00000397587.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
ENST00000409078.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
ENST00000455322.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
ENST00000541377.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
ENST00000541859.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
ENST00000545861.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
NM_001142273.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
NM_001142274.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
NM_001207051.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036430.hg.1 |
NM_015282.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000263710.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000397587.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000409078.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000452274.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000455322.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000463621.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000541377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_001142273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_001142274.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_001207051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_015282.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000263710.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000397587.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000409078.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000452274.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000455322.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000463621.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
ENST00000541377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_001142273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_001142274.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_001207051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
NM_015282.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203090.hg.1 |
TC0200014091.hg.1 |
Clariom D TC |