Assay Details
| Gene Symbol: UBXN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015562.1 | NP_056377.1 | 5 - 6 | 537 | 65 | |
| XM_011512670.2 | XP_011510972.1 | 5 - 6 | 485 | 65 | ||
| XM_011512671.2 | XP_011510973.1 | 4 - 5 | 342 | 65 | ||
| GenBank mRNA | AB018337.1 | - | 5 - 6 | 465 | 65 | |
| AK026451.1 | - | 5 - 6 | 490 | 65 | ||
| AK129880.1 | - | 5 - 6 | 490 | 65 | ||
| AK303630.1 | - | 4 - 5 | 342 | 65 | ||
| BC028986.1 | - | 4 - 5 | 369 | 65 | ||
| BC047420.1 | - | 5 - 6 | 487 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002159.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK303630.1,XM_011512671.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000383917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | ENST00000495872.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038243.hg.1 | TC03002159.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038243.hg.1 | ENST00000296328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038243.hg.1 | ENST00000535858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038243.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038242.hg.1 | TC03002159.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038242.hg.1 | ENST00000296328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038242.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038242.hg.1 | ENST00000535858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038242.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202407.hg.1 | TC0300013701.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000381887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000383917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000413584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000428095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000493566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | ENST00000495872.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013701.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202409.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202409.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202409.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202409.hg.1 | ENST00000296328.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202409.hg.1 | ENST00000429160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202409.hg.1 | NM_015562.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202409.hg.1 | TC0300013701.hg.1 | Clariom D TC |

UBXN7
Genomic Map