Assay Details
| Gene Symbol: DTX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300727.1 | NP_001287656.1 | 4 - 5 | 1010 | 84 | |
| NM_015177.1 | NP_055992.1 | 4 - 5 | 1419 | 84 | ||
| XM_006718483.3 | XP_006718546.1 | 4 - 5 | 1463 | 84 | ||
| GenBank mRNA | AB023154.1 | - | 4 - 5 | 1266 | 84 | |
| BC119011.1 | - | 4 - 5 | 1010 | 40085251 | 84 | |
| BC122861.1 | - | 4 - 5 | 1010 | 40085252 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000484.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11005884.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005884.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005884.hg.1 | ENST00000532982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005884.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005884.hg.1 | TC11000484.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000527475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007729.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | ENST00000227451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | ENST00000531902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | ENST00000532982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | NM_001300727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | NM_015177.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154486.hg.1 | TC1100007729.hg.1 | Clariom D TC |

DTX4
Genomic Map