Assay Details
| Gene Symbol: FRMD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015123.2 | NP_055938.1 | 6 - 7 | 870 | 111 | |
| XM_005264722.1 | XP_005264779.1 | 6 - 7 | 547 | 111 | ||
| XM_017005989.1 | XP_016861478.1 | 6 - 7 | 654 | 111 | ||
| XM_017005990.1 | XP_016861479.1 | 6 - 7 | 547 | 111 | ||
| XM_017005991.1 | XP_016861480.1 | 7 - 8 | 707 | 111 | ||
| XM_017005992.1 | XP_016861481.1 | 6 - 7 | 487 | 111 | ||
| XM_017005993.1 | XP_016861482.1 | 6 - 7 | 478 | 111 | ||
| XM_017005994.1 | XP_016861483.1 | 6 - 7 | 2320 | 111 | ||
| GenBank mRNA | AB023230.1 | - | 6 - 7 | 639 | 111 | |
| AK000244.1 | - | 6 - 7 | 451 | 111 | ||
| AK295045.1 | - | 1 - 2 | 112 | 111 | ||
| DC340455.1 | - | 1 - 2 | 81 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

FRMD4B
Genomic Map