Assay Details
| Gene Symbol: INTU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015693.3 | NP_056508.2 | 9 - 10 | 1607 | 60 | |
| XM_011531844.2 | XP_011530146.1 | 9 - 10 | 1631 | 60 | ||
| XM_011531846.1 | XP_011530148.1 | 9 - 10 | 1579 | 60 | ||
| XM_011531847.1 | XP_011530149.1 | 9 - 10 | 1016 | 60 | ||
| XM_011531848.2 | XP_011530150.1 | 4 - 5 | 442 | 60 | ||
| XM_011531849.2 | XP_011530151.1 | 9 - 10 | 1634 | 60 | ||
| XM_017008025.1 | XP_016863514.1 | 6 - 7 | 550 | 60 | ||
| GenBank mRNA | AB033110.1 | - | 9 - 10 | 1574 | 60 | |
| AK123320.1 | - | 2 - 3 | 947 | 60 | ||
| AK291481.1 | - | 9 - 10 | 1589 | 60 | ||
| BC051698.1 | - | 11 - 12 | 4075 | 60 | ||
| BC130611.1 | - | 9 - 10 | 1573 | 40146642 | 60 | |
More Information
Set Membership: |
|

INTU
Genomic Map