Assay Details
| Gene Symbol: TNRC6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018996.3 | NP_061869.2 | 17 - 18 | 4907 | 65 | |
| XM_006722000.3 | XP_006722063.3 | 18 - 19 | 10550 | 65 | ||
| XM_017024907.1 | XP_016880396.1 | 18 - 19 | 10559 | 65 | ||
| GenBank mRNA | AB046802.2 | - | 14 - 15 | 4457 | 65 | |
| AL834429.1 | - | 14 - 15 | 2429 | 65 | ||
More Information
Set Membership: |
|

TNRC6C
Genomic Map