Assay Details
| Gene Symbol: TSC22D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183422.3 | NP_904358.2 | 1 - 2 | 3405 | 94 | |
| GenBank mRNA | AB082525.1 | - | 1 - 2 | 3405 | 94 | |
| AK027071.1 | - | 1 - 2 | 1960 | 94 | ||
| BC136475.1 | - | 1 - 2 | 3175 | 9020464 | 94 | |
| BC136482.1 | - | 1 - 2 | 3175 | 9020471 | 94 | |
| CR627459.1 | - | 2 - 3 | 1598 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000622.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000261489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000437455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000437455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000437455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000458659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000460842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000472477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000486464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000487881.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000493016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000493016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000493016.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000496838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000501704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000611198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | ENST00000622051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_001243799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_006022.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008793.hg.1 | NM_183422.3 | Refseq |

TSC22D1
Genomic Map