Assay Details
| Gene Symbol: ESRRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004452.3 | NP_004443.3 | 2 - 3 | 308 | 73 | |
| XM_011536550.2 | XP_011534852.1 | 2 - 3 | 544 | 73 | ||
| XM_017021085.1 | XP_016876574.1 | 1 - 2 | 115 | 73 | ||
| XM_017021086.1 | XP_016876575.1 | 2 - 3 | 545 | 73 | ||
| GenBank mRNA | DR007211.1 | - | 2 - 3 | 315 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000481.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_004452.3,XM_011536550.2,XM_017021085.1,XM_017021086.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
embryonic placenta development
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
intracellular receptor signaling pathway
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II distal enhancer sequence-specific DNA binding
steroid hormone receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
transcription factor binding
zinc ion binding
sequence-specific DNA binding
embryonic placenta development
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
intracellular receptor signaling pathway
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II distal enhancer sequence-specific DNA binding
steroid hormone receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
transcription factor binding
zinc ion binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14006084.hg.1 | ENST00000261532.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006084.hg.1 | ENST00000380887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006084.hg.1 | ENST00000507951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006084.hg.1 | ENST00000509242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006084.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006084.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006084.hg.1 | TC14000481.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000509323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000509323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | ENST00000611036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007725.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000380887.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000505752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000507951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000509242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000512784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | ENST00000556177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | NM_004452.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154826.hg.1 | TC1400007725.hg.1 | Clariom D TC |

ESRRB
Genomic Map