Assay Details
| Gene Symbol: PAPPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020318.2 | NP_064714.2 | 18 - 19 | 6045 | 70 | |
| XM_005245422.3 | XP_005245479.1 | 18 - 19 | 6092 | 70 | ||
| XM_011509857.1 | XP_011508159.1 | 19 - 20 | 5962 | 70 | ||
| XM_011509858.2 | XP_011508160.1 | 18 - 19 | 6305 | 70 | ||
| XM_017002023.1 | XP_016857512.1 | 19 - 20 | 6143 | 70 | ||
| XM_017002024.1 | XP_016857513.1 | 19 - 20 | 6190 | 70 | ||
| XM_017002025.1 | XP_016857514.1 | 18 - 19 | 6296 | 70 | ||
| GenBank mRNA | AB209369.1 | - | 10 - 11 | 1583 | 70 | |
| AF311940.1 | - | 17 - 18 | 4881 | 70 | ||
| AF342989.1 | - | 18 - 19 | 6045 | 70 | ||
| AJ278348.1 | - | 17 - 18 | 4844 | 70 | ||
| BC152552.1 | - | 18 - 19 | 6033 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001534.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01023677.hg.1 | TC01001534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023677.hg.1 | ENST00000367662.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023677.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178713.hg.1 | ENST00000367662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178713.hg.1 | NM_020318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178713.hg.1 | TC0100010713.hg.1 | Clariom D TC |

PAPPA2
Genomic Map