Assay Details
| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF469199.1 | - | 1 - 2 | 152 | 111 | |
| AF469200.1 | - | 1 - 2 | 152 | 111 | ||
| BC111049.1 | - | 1 - 2 | 171 | 111 | ||
More Information
Set Membership: |
|

DIO3OS
Genomic Map