Assay Details
| Gene Symbol: B4GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159387.1 | NP_001152859.1 | 4 - 5 | 480 | 69 | |
| NM_001159388.1 | NP_001152860.1 | 4 - 5 | 493 | 69 | ||
| NM_153446.2 | NP_703147.2 | 4 - 5 | 695 | 69 | ||
| XM_011524314.2 | XP_011522616.1 | 4 - 5 | 848 | 69 | ||
| XM_017024173.1 | XP_016879662.1 | 4 - 5 | 542 | 69 | ||
| GenBank mRNA | AF510036.1 | - | 4 - 5 | 695 | 69 | |
| AJ517770.1 | - | 4 - 5 | 695 | 69 | ||
| AJ517771.1 | - | 4 - 5 | 477 | 69 | ||
| AK302876.1 | - | 4 - 5 | 493 | 69 | ||
| BC113675.1 | - | 4 - 5 | 689 | 8322727 | 69 | |
| BC113677.1 | - | 4 - 5 | 689 | 8322729 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000638.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17007975.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007975.hg.1 | ENST00000393354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007975.hg.1 | ENST00000504681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007975.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007975.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007975.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007975.hg.1 | TC17000638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160591.hg.1 | TC1700008191.hg.1 | Clariom D TC |

B4GALNT2
Genomic Map