Assay Details
| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080492.1 | NP_001073961.1 | 2 - 3 | 391 | 76 | |
| NM_001305173.1 | NP_001292102.1 | 2 - 3 | 388 | 76 | ||
| NM_001305174.1 | NP_001292103.1 | 2 - 3 | 391 | 76 | ||
| GenBank mRNA | AK058068.1 | - | 2 - 3 | 333 | 76 | |
| AK302175.1 | - | 2 - 3 | 321 | 76 | ||
| BC057843.1 | - | 2 - 3 | 380 | 5296090 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001151.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK058068.1,AK302175.1,BC057843.1,NM_001080492.1,NM_001305173.1,NM_001305174.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016026.hg.1 | ENST00000219301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016026.hg.1 | ENST00000543437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016026.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016026.hg.1 | TC16001151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016025.hg.1 | ENST00000219301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016025.hg.1 | ENST00000543437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016025.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016025.hg.1 | TC16001151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016028.hg.1 | ENST00000219301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016028.hg.1 | ENST00000543437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016028.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016028.hg.1 | TC16001151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016029.hg.1 | ENST00000219301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016029.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016029.hg.1 | TC16001151.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180593.hg.1 | TC1600010480.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |

PRSS54
Genomic Map