Assay Details
| Gene Symbol: SRRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110199.2 | NP_001103669.1 | 14 - 15 | 1951 | 71 | |
| GenBank mRNA | AK092590.1 | - | 2 - 3 | 369 | 71 | |
| BE466083.1 | - | 3 - 4 | 437 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000483.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK092590.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006756.hg.1 | TC07000483.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006756.hg.1 | ENST00000326382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006756.hg.1 | ENST00000388802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006756.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156078.hg.1 | TC0700008088.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000464752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000479294.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000611745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | ENST00000612155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001110199.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008088.hg.1 | NM_001291831.1 | Refseq |

SRRM3
Genomic Map