Assay Details
| Gene Symbol: TRMT44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152544.2 | NP_689757.2 | 8 - 9 | 1518 | 65 | |
| XM_005247950.3 | XP_005248007.1 | 8 - 9 | 1515 | 65 | ||
| XM_011513406.2 | XP_011511708.1 | 8 - 9 | 1547 | 65 | ||
| XM_011513407.2 | XP_011511709.1 | 8 - 9 | 1547 | 65 | ||
| XM_011513408.1 | XP_011511710.1 | 9 - 10 | 1659 | 65 | ||
| XM_011513409.2 | XP_011511711.1 | 9 - 10 | 1630 | 65 | ||
| GenBank mRNA | AK093044.1 | - | 4 - 5 | 524 | 65 | |
| BC035655.1 | - | 8 - 9 | 1018 | 5588410 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000090.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04001894.hg.1 | TC04000090.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001894.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001894.hg.1 | ENST00000513449.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001894.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000504134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000504134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000528167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000528167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000528167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000531038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000531038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000531038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400146872.hg.1 | TC0400006756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400146873.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146873.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146873.hg.1 | TC0400006756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000389737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000504134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000504134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000511366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000513449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000528167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000528167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000528167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000531038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000531038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000531038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | ENST00000532937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006756.hg.1 | NM_152544.2 | Refseq |

TRMT44
Genomic Map