Assay Details
| Gene Symbol: DDX51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175066.3 | NP_778236.2 | 11 - 12 | 1712 | 101 | |
| GenBank mRNA | AB593157.1 | - | 11 - 12 | 1732 | 101 | |
| AK097078.1 | - | 11 - 12 | 1712 | 101 | ||
| BC012461.2 | - | 2 - 3 | 235 | 101 | ||
| BC040185.2 | - | 11 - 12 | 1685 | 4801253 | 101 | |
| CR936870.1 | - | 3 - 4 | 731 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002148.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028122.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028122.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028122.hg.1 | TC12002148.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197634.hg.1 | TC1200012512.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 164685 | - | - |

DDX51
Genomic Map