Assay Details
| Gene Symbol: HCCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122608.2 | NP_001116080.1 | 4 - 5 | 619 | 128 | |
| NM_001171991.2 | NP_001165462.1 | 4 - 5 | 582 | 128 | ||
| NM_005333.4 | NP_005324.3 | 4 - 5 | 677 | 128 | ||
| GenBank mRNA | AK097815.1 | - | 4 - 5 | 604 | 128 | |
| BC001691.2 | - | 4 - 5 | 526 | 3030501 | 128 | |
| BC095455.1 | - | 4 - 5 | 620 | 30512178 | 128 | |
| CR749578.1 | - | 4 - 5 | 634 | 128 | ||
| U36787.1 | - | 4 - 5 | 601 | 128 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000042.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000559.hg.1 | TC0X000042.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000559.hg.1 | ENST00000321143.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000559.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000559.hg.1 | ENST00000380763.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000559.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000559.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000559.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145140.hg.1 | TC0X00006603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000321143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | ENST00000380763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001122608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_001171991.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006603.hg.1 | NM_005333.4 | Refseq |

HCCS
Genomic Map