Assay Details
| Gene Symbol: TNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022093.1 | NP_071376.1 | 9 - 10 | 2234 | 76 | |
| XM_017002048.1 | XP_016857537.1 | 9 - 10 | 2239 | 76 | ||
| XM_017002049.1 | XP_016857538.1 | 9 - 10 | 2240 | 76 | ||
| GenBank mRNA | AK127044.1 | - | 9 - 10 | 2234 | 76 | |
| AL049689.1 | - | 8 - 9 | 2106 | 76 | ||
| BC136619.1 | - | 8 - 9 | 2151 | 9020609 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001527.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01023621.hg.1 | TC01001527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023621.hg.1 | ENST00000239462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023621.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023623.hg.1 | TC01001527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023623.hg.1 | ENST00000239462.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023623.hg.1 | ENST00000539081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023623.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178586.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178586.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178586.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178586.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178586.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178586.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178586.hg.1 | TC0100010689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178588.hg.1 | TC0100010689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |

TNN
Genomic Map