Assay Details
| Gene Symbol: DKFZP586I1420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002186.1 | 1 | 547 | 141 | ||
| GenBank mRNA | AL050378.1 | - | 1 | 547 | 141 | |
| BC073840.1 | - | 1 | 731 | 141 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc and ring finger 2, E3 ubiquitin protein ligase |
ZNRF2 | RNF202 | -2.4kb | Sense | Chr. 7 - 30284307 - 30367692 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1176 | LINC01176 | 18.1kb | Sense | Chr. 7 - 30390885 - 30404055 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000196.hg.1
Set Membership: |
|

DKFZP586I1420
Genomic Map