Assay Details
| Gene Symbol: ELMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206480.2 | NP_001193409.1 | 3 - 4 | 653 | 104 | |
| NM_001206482.1 | NP_001193411.1 | 3 - 4 | 823 | 104 | ||
| NM_014800.10 | NP_055615.8 | 3 - 4 | 464 | 104 | ||
| XM_005249919.2 | XP_005249976.1 | 4 - 5 | 1017 | 104 | ||
| XM_006715805.1 | XP_006715868.1 | 3 - 4 | 309 | 104 | ||
| XM_011515654.1 | XP_011513956.1 | 4 - 5 | 1702 | 104 | ||
| XM_017012838.1 | XP_016868327.1 | 4 - 5 | 1887 | 104 | ||
| XM_017012839.1 | XP_016868328.1 | 3 - 4 | 251 | 104 | ||
| GenBank mRNA | AF398885.1 | - | 2 - 3 | 115 | 104 | |
| AK001084.1 | - | 3 - 4 | 369 | 104 | ||
| AK291784.1 | - | 3 - 4 | 823 | 104 | ||
| AK314349.1 | - | 3 - 4 | 253 | 104 | ||
| AL136787.1 | - | 3 - 4 | 365 | 104 | ||
| BC114341.2 | - | 3 - 4 | 202 | 40035461 | 104 | |
| DB156714.1 | - | 3 - 4 | 420 | 104 | ||
More Information
Set Membership: |
|

ELMO1
Genomic Map