Assay Details
| Gene Symbol: FILIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300866.1 | NP_001287795.1 | 6 - 7 | 3911 | 106 | |
| GenBank mRNA | AL832009.2 | - | 6 - 7 | 3711 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

FILIP1
Genomic Map