Assay Details
| Gene Symbol: TMEM245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032012.3 | NP_114401.2 | 13 - 14 | 2006 | 91 | |
| XM_011518446.2 | XP_011516748.1 | 13 - 14 | 2021 | 91 | ||
| XM_011518447.1 | XP_011516749.1 | 12 - 13 | 1982 | 91 | ||
| XM_011518449.1 | XP_011516751.1 | 12 - 13 | 1889 | 91 | ||
| XM_011518451.1 | XP_011516753.1 | 11 - 12 | 1877 | 91 | ||
| XM_011518452.1 | XP_011516754.1 | 12 - 13 | 1772 | 91 | ||
| XM_017014571.1 | XP_016870060.1 | 12 - 13 | 1979 | 91 | ||
| XM_017014572.1 | XP_016870061.1 | 11 - 12 | 1748 | 91 | ||
| GenBank mRNA | AF153415.1 | - | 13 - 14 | 2006 | 91 | |
| AK172815.1 | - | 7 - 8 | 751 | 91 | ||
| AK299952.1 | - | 13 - 14 | 1988 | 91 | ||
| AL556162.3 | - | 7 - 8 | 717 | 91 | ||
| AL833960.1 | - | 11 - 12 | 1397 | 91 | ||
| BC014248.2 | - | 3 - 4 | 357 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001459.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018070.hg.1 | TC09001459.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018070.hg.1 | ENST00000223608.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018070.hg.1 | ENST00000374586.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018070.hg.1 | ENST00000374587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018070.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175856.hg.1 | TC0900011151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000374586.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000413712.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000472207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000482868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | ENST00000491854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011151.hg.1 | NM_032012.3 | Refseq |

TMEM245
Genomic Map