Assay Details
Gene Symbol: COG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153603.3 | NP_705831.1 | 15 - 16 | 2195 | 56 | |
XM_017023870.1 | XP_016879359.1 | 15 - 16 | 2238 | 56 | ||
GenBank mRNA | AF070568.1 | - | 7 - 8 | 767 | 56 | |
AK093989.1 | - | 12 - 13 | 1551 | 56 | ||
AK130714.1 | - | 2 - 3 | 1980 | 56 | ||
AY358632.1 | - | 10 - 11 | 1232 | 56 | ||
BC000549.2 | - | 15 - 16 | 2150 | 3163316 | 56 | |
BC037563.1 | - | 15 - 16 | 2184 | 5393265 | 56 | |
KU178837.1 | - | 15 - 16 | 1998 | 56 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000960.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000569635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | ENST00000569635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16013832.hg.1 | ENST00000307149.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16013832.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16013832.hg.1 | TC16000960.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600174901.hg.1 | TC1600009731.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000307149.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000561854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000563164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000566364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000567821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000569635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | ENST00000569635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009731.hg.1 | NM_153603.3 | Refseq |
